Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-2,3-Epoxy-1-propanol
(±)-2,3-Epoxy-1-propanol, Sigma-Aldrich®
(±)-2,3-Epoxy-1-propanol, Sigma-Aldrich®
(±)-2,3-Epoxy-1-propanol, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
(±)-2,3-Epoxy-1-propanol, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
556-52-5
:
Formula:
C₃H₆O₂
MDL Number:
MFCD00005147
ADR:
6.1,III
:
M-Clarity™ quality level = MQ200
:
74.08 g/mol
EINECS:
209-128-3
UN:
2810