Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Palladium
Palladium 10% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Palladium 10% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Palladium 10% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Palladium 10% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
7440-05-3
:
Formula:
Pd
MDL Number:
MFCD00011167
:
106.420 g/mol
EINECS:
231-115-6
ADR:
4.1,III