0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
(S)-(-)-2,3-Epoxy-1-propanol
(S)-(-)-2,3-Epoxy-1-propanol, Sigma-Aldrich®
(S)-(-)-2,3-Epoxy-1-propanol, Sigma-Aldrich®
(S)-(-)-2,3-Epoxy-1-propanol, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
(S)-(-)-2,3-Epoxy-1-propanol, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
60456-23-7
:
Formula:
C₃H₆O₂
MDL Number:
MFCD00074874
:
74.08 g/mol
:
M-Clarity™ quality level = MQ100