0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Cysteaminium chloride
Cysteaminium chloride, Sigma-Aldrich®
Cysteaminium chloride, Sigma-Aldrich®
Cysteaminium chloride, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Cysteaminium chloride, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
156-57-0
:
Formula:
C₂H₇NS·HCl
MDL Number:
MFCD00012904
:
113.61 g/mol
:
M-Clarity™ quality level = MQ200
EINECS:
205-858-1