Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-2,3-Epoxybutane
cis-2,3-Epoxybutane, Sigma-Aldrich®
cis-2,3-Epoxybutane, Sigma-Aldrich®
SIAL294047-1G
:
Merck
:
cis-2,3-Epoxybutane, Sigma-Aldrich®
SIAL294047-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
72.11 g/mol
:
3,II
:
M-Clarity™ quality level = MQ100
:
3271
:
1758-33-4
:
C₄H₈O
:
MFCD00005128