Disodium benzene-1,3-disulphonate, Sigma-Aldrich®
Catalog # SIAL259802-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:282.21 g/mol
:M-Clarity™ quality level = MQ100
EINECS:212-606-4
CAS Number:831-59-4
Formula:C₆H₄Na₂O₆S₂
MDL Number:MFCD00007492