1,3-Dichloro-5,5-dimethylhydantoin, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:118-52-5
:
Formula:C₅H₆Cl₂N₂O₂
MDL Number:MFCD00003190
:197.02 g/mol
ADR:5.1,II
:M-Clarity™ quality level = MQ200
EINECS:204-258-7
UN:1479