Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thiophosphoryl chloride
Thiophosphoryl chloride, Sigma-Aldrich®
Thiophosphoryl chloride, Sigma-Aldrich®
Thiophosphoryl chloride, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Thiophosphoryl chloride, Sigma-Aldrich®
Supplier:
Merck
Certificates
About this item
CAS Number:
3982-91-0
:
Formula:
Cl₃PS
MDL Number:
MFCD00011504
:
169.4 g/mol
ADR:
8,II
:
M-Clarity™ quality level = MQ200
EINECS:
223-622-6
UN:
1837
Related Information
Documentation
Certificates
Search by Lot Number
Documentation