Palladium 10% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
ADR:4.1,III
:M-Clarity™ quality level = MQ200
EINECS:231-115-6