Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Diacryloylpiperazine
1,4-Diacryloylpiperazine, Sigma-Aldrich®
1,4-Diacryloylpiperazine, Sigma-Aldrich®
1,4-Diacryloylpiperazine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
1,4-Diacryloylpiperazine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
6342-17-2
:
Formula:
C₁₀H₁₄N₂O₂
MDL Number:
MFCD00077661
:
194.23 g/mol