N-tert-Butylbenzylamine
MOLE61781090-100G
: Molekula
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
: N-Benzyl-tert-butylamine, N-Benzyl-2-methylpropan-2-amine
:
:163.26 g/mol
:222-179-6
:2735
:3378-72-1
:C₁₁H₁₇N
:MFCD00008798
:8 III