1,4-Diacryloylpiperazine
Catalog # MOLEM64913439
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:194.23 g/mol
CAS Number:6342-17-2
Formula:C₁₀H₁₄N₂O₂
MDL Number:MFCD00077661