(S)-(+)-2-Chloromandelic acid
Glass bottle for solids, 500 mg
Catalog # MOLEM45255220
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: (S)-2-(2-Chlorophenyl)-2-hydroxyacetic acid
- CAS Number:52950-19-3
- Formula:C₈H₇ClO₃
- MDL Number:MFCD00798437
- Molecular Weight:186.59 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



