(S)-(-)-Hydroxypyrrolidine
Glass bottle for solids, 1 g
Catalog # MOLEM43186047
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: (S)-(-)-3-Pyrrolidinol, (S)-Pyrrolidin-3-ol, (S)-(-)-3-Hydroxypyrrolidine
- CAS Number:100243-39-8
- Formula:C₄H₉NO
- MDL Number:MFCD00192426
- Molecular Weight:87.12 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



