Order Entry
Export
ContactUsLinkComponent
Benzyl chloroformate
Corrosive
Environment
Benzyl chloroformate
Catalog # MOLE28624136
Supplier:  Molekula
CAS Number:  
Corrosive
Environment
Benzyl chloroformate
Catalog # MOLE28624136
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :170.6 g/mol
EINECS:207-925-0
UN:1739
ADR:8,I
 :GHS05
 :GHS09
CAS Number:501-53-1
Formula:C₈H₇ClO₂
MDL Number:MFCD00000640