(S)-(+)-β-Aminophenethyl alcohol
Glass bottle for solids, 5 g
Catalog # MOLEM25963926
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol
- CAS Number:20989-17-7
- Formula:C₈H₁₁NO
- MDL Number:MFCD00064404
- Molecular Weight:137.18 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



