Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-(+)-Hydroxypyrrolidine
(R)-(+)-Hydroxypyrrolidine
(R)-(+)-Hydroxypyrrolidine
(R)-(+)-Hydroxypyrrolidine
MOLEM15970045
:
Molekula
:
(R)-(+)-Hydroxypyrrolidine
MOLEM15970045
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
87.12 g/mol
:
2799-21-5
:
C₄H₉NO
:
MFCD00145220