Organic reference standard, Acrolein 100 µg/ml in acetone, Dr Ehrenstorfer
Catalog # EHERXA10045000AC
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:107-02-8
- Description:Acrolein 100 µg/ml in acetone
- Formula:C₃H₄O
- Description:acrolein