
Aluminium chloride, anhydrous ≥99.985% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7446-70-0
:
Formula:AlCl₃
MDL Number:MFCD00003422
:133.34 g/mol
Merck Index:14,337
Boiling Pt:180 °C (1013 hPa)
Melting Pt:190 °C (subl.)
Density:2.44
Storage Temperature:Ambient
EINECS:231-208-1
UN:1726
ADR:8,II
:GHS05