6-Hydroxypicolinic acid
Glass bottle for solids, 25 g
Catalog # COBBCA-4145-25G
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
Synonyms: 6-Hydroxy-2-pyridinecarboxylic acid, 6-Hydroxypyridine-2-carboxylic acid
- CAS Number:19621-92-2
- Formula:C₆H₅NO₃
- MDL Number:MFCD00192220
- Molecular Weight:139.11 g/mol
- EINECS:417-690-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



