Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-2,3-Epoxybutane
cis-2,3-Epoxybutane ≥98%
cis-2,3-Epoxybutane ≥98%
cis-2,3-Epoxybutane ≥98%
L10320.06
:
Thermo Fisher Scientific
:
cis-2,3-Epoxybutane ≥98%
L10320.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
1758-33-4
:
:
C₄H₈O
:
MFCD00005128
:
72.11 g/mol
:
60…61 °C
:
–84 °C
:
0.826
:
–21 °C (–5 °F)
:
3271
:
3,II
:
GHS02
:
GHS08
:
GHS07
:
Ambient