1-Iodo-2-(trimethylsilyl)acetylene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18163-47-8
:
Formula:C₅H₉ISi
MDL Number:MFCD00274201
Boiling Pt:130 °C
Density:1.46
Flash Pt:48 °C (118 °F)
ADR:3,III
:224.12 g/mol
:GHS08
:GHS02
:GHS07
Storage Temperature:Ambient