Diethyl phenylmalonate ≥98%
Catalog # B20234.30
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Environmentally Preferable:
Specifications
About this item
Synonyms: Diethyl 2-phenylmalonate
CAS Number:83-13-6
:
Formula:C₁₃H₁₆O₄
MDL Number:MFCD00009144
:236.27 g/mol
Boiling Pt:170…172 °C (14 mmHg)
Melting Pt:15…16 °C
Density:1.096
Flash Pt:120 °C (248 °F)
Storage Temperature:Ambient
EINECS:201-456-5