Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium 2,4-dinitrobenzenesulphonate
Sodium-2,4-dinitrobenzenesulphonate ≥97%
Sodium-2,4-dinitrobenzenesulphonate ≥97%
:
Thermo Fisher Scientific
Sodium-2,4-dinitrobenzenesulphonate ≥97%
:
Thermo Fisher Scientific
:
885-62-1
:
:
C₆H₃N₂NaO₇S
:
MFCD00007471
:
270.15 g/mol
:
212-943-7
:
GHS07
:
Ambient
msds_ALFAA18493
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years