

Benzyl chloroformate 95% stabilised
Supplier: abcr
Certificates
About this item
CAS Number:501-53-1
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00000640
:170.6 g/mol
Merck Index:14,1801
Boiling Pt:178…180 °C
Melting Pt:–20 °C
Density:1.195
Flash Pt:125 °C (257 °F)
UN:1739
ADR:8,I
:Stabilised with sodium carbonate (Na₂CO₃)
EINECS:207-925-0
:GHS06
:GHS05
Storage Temperature:Freezer