4'-Ethoxyacetanilide ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 62-44-2 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 134-136° Beilstein Reference: 1869238 Merck Reference: 14,7204 MDL No.: MFCD00009094
Synonyms: Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide
- CAS Number:62-44-2
- Formula:C₁₀H₁₃NO₂
- MDL Number:MFCD00009094
- Molecular Weight:179.22 g/mol
- Merck Index:14,7204
- Melting Pt:134…136 °C
- Storage Temperature:Ambient
- EINECS:200-533-0
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 62-44-2 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 134-136° Beilstein Reference: 1869238 Merck Reference: 14,7204 MDL No.: MFCD00009094
Synonyms: Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide
- CAS Number:62-44-2
- Formula:C₁₀H₁₃NO₂
- MDL Number:MFCD00009094
- Molecular Weight:179.22 g/mol
- Merck Index:14,7204
- Melting Pt:134…136 °C
- Storage Temperature:Ambient
- EINECS:200-533-0



