
Bis(trimethylsilyl)acetylene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14630-40-1
:
Formula:C₈H₁₈Si₂
MDL Number:MFCD00008276
:170.4 g/mol
Boiling Pt:136…137 °C (760 mmHg)
Melting Pt:21…24 °C
Density:0.75 g/cm³
Flash Pt:2 °C
Storage Temperature:Refrigerator
EINECS:238-671-9
UN:1325
ADR:4.1,II
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.4260 to 1.4280 (20°C, 589 nm)
- GC:≥98.5 %
- HPLC:≥97.5 %