



1,3-Dibromo-5,5-dimethylhydantoin 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-48-5
:
Formula:C₅H₆Br₂N₂O₂
MDL Number:MFCD00003189
Melting Pt:187…191 °C (dec.)
Flash Pt:155 °C
Storage Temperature:Ambient
EINECS:201-030-9
UN:3085
ADR:5.1,II
:285.92 g/mol
:GHS07
:GHS05
:GHS09
:GHS03
Specification Test Results
- Appearance:Cream to light yellow Powder
- Infrared spectrum:Conforms
- Titration Iodometric:≥97.5 %
- Loss on drying:≤0.5 %