



Benzyl chloroformate 97% (w/w) stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:501-53-1
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00000640
:170.6 g/mol
Boiling Pt:178…180 °C (760 mmHg)
Melting Pt:–20 °C
Density:1.2 g/cm³
Flash Pt:92 °C
EINECS:207-925-0
UN:1739
ADR:8,I
:GHS07
:GHS08
:GHS05
:GHS09
Storage Temperature:Freezer
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5160 to 1.5180 (20°C, 589 nm)
- Additional info:important note: the material builds progressively benzyl chloride under normal storage conditions
- GC:≥96.0 %
- Color scale:≤100 APHA
- Stabilizer:0.1 % anhydrous sodium carbonate
- Phosgene:≤0.1 %
- Iron (Fe):≤2 ppm
- Acidity (HCl):≤0.3 %
- Benzyl alcohol:≤0.5 %