Order Entry
Export
ContactUsLinkComponent
 
 
 
(±)-2,3-Epoxy-1-propanol 96%
 
undefined
 
 
(±)-2,3-Epoxy-1-propanol 96%

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4300 to 1.4360 (20°C, 589 nm)
  • GC:
    ≥95 %