


Sodium metavanadate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13718-26-8
:
Formula:NaO₃V
MDL Number:MFCD00011125
:121.93 g/mol
Merck Index:12,08849
Melting Pt:630 °C
Density:5,15 g/cm³ (20 °C)
EINECS:237-272-7
UN:3285
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to light beige to light green Powder
- Titration with Na2S2O3:≥95 %