Sulphuric acid 95,0 - 100,5% 95.0-100.5% Ph. Eur.
:7664-93-9
:
:H₂SO₄
:MFCD00064589
:330 °C (1013 hPa)
:10.38 °C
:1.84 g/cm³ (20 °C)
:98.08 g/mol
:Ambient
:231-639-5
:1830
:8 II
:01-2119458838-20
:GHS05
- Environmentally Preferable:
- Assay:95.0 - 100.5 %
- Appearance:Clear viscous liquid
- Identification A:Passes test
- Identification (SO4):Passes test
- Appearance of solution:Passes test
- Cl (Chloride):≤ 50 ppm
- NO3 (Nitrate):Passes test
- Fe (Iron):≤ 25 ppm
- Heavy metals (as Pb):≤ 5 ppm
- Residual solvents:Passes test
- Residual metal catalysts (skip lot)::.
- As (Arsenic):≤ 0.1 ppm
- Cd (Cadmium):≤ 0.1 ppm
- Co (Cobalt):≤ 0.1 ppm
- Hg (Mercury):≤ 0.1 ppm
- Ni (Nickel):≤ 0.1 ppm
- Pb (Lead):≤ 0.1 ppm
- V (Vanadium):≤ 0.1 ppm
- Conforms to Ph.Eur.:Passes test