Sodium sulphite, anhydrous 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7757-83-7
:
Formula:Na₂SO₃
MDL Number:MFCD00003503
:126.04 g/mol
Merck Index:14,8682
Melting Pt:>500 °C (dec.)
Density:2.633
Storage Temperature:Ambient
EINECS:231-821-4
REACH:01-2119537420-49