
Sodium thiocyanate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:540-72-7
:
Formula:NaSCN
MDL Number:MFCD00011123
:81.07 g/mol
Merck Index:14,9327
Melting Pt:287 °C
Density:1.74
EINECS:208-754-4
:GHS07
Storage Temperature:Ambient