
Sodium thiocyanate 98+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:540-72-7
:
Formula:NaSCN
MDL Number:MFCD00011123
:81.07 g/mol
Merck Index:13,09401
Melting Pt:287 °C
Density:1,735 g/cm³ (20 °C)
EINECS:208-754-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder or crystals and/or chunks
- Titration Argentometric:≥98.0 %
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):≤100 ppm
- Ammonium (NH4):≤20 ppm
- Iron (Fe):≤2 ppm
- Sulfur (S):≤10 ppm
- Heavy metals (as Pb):≤5 ppm
- Impurity:≤0.2 % Carbonate (as Na2CO3)
- Insoluble matter:≤0.005 %