
Sodium thiocyanate 98%, extra pure
:540-72-7
:
:NaSCN
:MFCD00011123
:81.07 g/mol
:13,09401
:287 °C
:1,735 g/cm³ (20 °C)
:208-754-4
:GHS07
:Ambient
- Appearance:White Crystalline powder or crystals and/or chunks
- Titration Argentometric:≥97.5 % (on dried substance)
- Loss on drying:≤2.5 %
- pH:5.5 to 7.5 (5 % at 20°C)
- Iron (Fe):≤2 ppm