Order Entry
Export
ContactUsLinkComponent
 
Disodium hydrogen phosphate 99+% ACS suitable for buffer solutions
 
undefined
Disodium hydrogen phosphate 99+% ACS suitable for buffer solutions

 

  • Appearance:
    White Powder or granular powder
  • Loss on drying:
    ≤0.2 % (105°C)
  • pH:
    8.7 to 9.3 (5 % solution at 25.0°C)
  • Titration excess HCl:
    ≥99.0 %
  • Chloride (Cl):
    ≤20 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Iron (Fe):
    ≤20 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Insoluble matter:
    ≤0.01 %