Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium peroxide 95%

Sodium peroxide 95%

Oxidizing
Corrosive
Sodium peroxide 95%
Certificates
Sodium peroxide 95%
Sodium peroxide 95%

3 Options Available Below

Product Details & Documents

Purity: 95% CAS No.: 1313-60-6 Molecular Formula: Na2O2 Formula Weight: 77.98 Melting Point: 460° dec. Merck Reference: 14,8655 MDL No.: MFCD00003497

Synonyms:  di-Sodium peroxide
  • CAS Number:1313-60-6
  • Formula:Na₂O₂
  • MDL Number:MFCD00003497
  • Molecular Weight:77.98 g/mol
  • Merck Index:14,8655
  • Melting Pt:460 °C (dec.)
  • Density:2.805
  • Storage Temperature:Ambient
  • EINECS:215-209-4
  • UN:1504
  • ADR:5.1,I
Product Family Options

Product Information