Orders LinkContactUsLinkComponent
Potassium chlorate ≥99%
Potassium chlorate ≥99%
Potassium chlorate ≥99%
Potassium chlorate ≥99%
Purity: 99+% CAS No.: 3811-04-9 Molecular Formula: KClO3 Formula Weight: 122.55 Odour: Odourless Melting Point: 368° Boiling Point: 400° dec. Merck Reference: 14,7620 UN No.: 1485 MDL No.: MFCD00011361
  •  :3811-04-9
  •  :KClO₃
  •  :MFCD00011361
  •  :122.55 g/mol
  •  :14,7620
  •  :400 °C (dec.)
  •  :368 °C
  •  :2.32
  •  :Ambient
  •  :223-289-7
  •  :1485
  •  :5.1,II
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    250 g
    1 * 250 g
    1 * 250 g - 36,40 €
    36,40 €
    1 * 250 g
       36,40 €
  • Glass bottle for solids
    1.000 g
    1 * 1.000 g
    1 * 1.000 g - 88,10 €
    88,10 €
    1 * 1.000 g
       88,10 €
  • Plastic bottle for solids
    5.000 g
    1 * 5.000 g
    1 * 5.000 g - 277,00 €
    277,00 €
    1 * 5.000 g
       277,00 €