Potassium chloride
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Potassiumchloride
- CAS Number:7447-40-7
- Formula:KCl
- MDL Number:MFCD00011360
- Molecular Weight:74.55 g/mol
- Merck Index:14,07621
- Boiling Pt:1420 °C (1013 hPa)
- Melting Pt:773 °C
- Density:1.98 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-211-8
4 Options Available Below
Product Details & Documents
Synonyms: Potassiumchloride
- CAS Number:7447-40-7
- Formula:KCl
- MDL Number:MFCD00011360
- Molecular Weight:74.55 g/mol
- Merck Index:14,07621
- Boiling Pt:1420 °C (1013 hPa)
- Melting Pt:773 °C
- Density:1.98 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-211-8



