
Sodium carbonate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:497-19-8
:
Formula:Na₂CO₃
MDL Number:MFCD00003494
:105.99 g/mol
Merck Index:14,08596
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:854 °C
Density:2.53 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07