Sodium carbonate
Glass bottle for solids, 1 kg
Catalog # MOLE13743314-1KG
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Calcined soda, Carbonic acid disodium salt
- CAS Number:497-19-8
- Formula:Na₂CO₃
- MDL Number:MFCD00003494
- Molecular Weight:105.99 g/mol
- Merck Index:14,08596
- Boiling Pt:1600 °C (1013 hPa)
- Melting Pt:854 °C
- Density:2.53 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-838-8
- REACH:01-2119485498-19
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



