Potassium bromide
Glass bottle for solids, 250 g
Catalog # ROTH9252.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:7758-02-3
- Formula:KBr
- MDL Number:MFCD00011358
- Molecular Weight:119 g/mol
- Merck Index:13,07701
- Boiling Pt:1435 °C (1013 hPa)
- Melting Pt:730 °C
- Density:2.75 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-830-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



