Cyclohexene ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 110-83-8 Molecular Formula: C6H10 Formula Weight: 82.15 Flash Point: -20°(-4°F) Refractive Index: 1.4460 Odour: Petrol-like Melting Point: -104° Boiling Point: 83-85° Beilstein Reference: 906737 Merck Reference: 14,2727 UN No.: 2256 MDL No.: MFCD00001539
- CAS Number:110-83-8
- Formula:C₆H₁₀
- MDL Number:MFCD00001539
- Molecular Weight:82.15 g/mol
- Merck Index:14,2727
- Boiling Pt:83…85 °C
- Melting Pt:–104 °C
- Density:0.811
- Flash Pt:–20 °C (–4 °F)
- Storage Temperature:Ambient
- EINECS:203-807-8
- UN:2256
- ADR:3,II
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 110-83-8 Molecular Formula: C6H10 Formula Weight: 82.15 Flash Point: -20°(-4°F) Refractive Index: 1.4460 Odour: Petrol-like Melting Point: -104° Boiling Point: 83-85° Beilstein Reference: 906737 Merck Reference: 14,2727 UN No.: 2256 MDL No.: MFCD00001539
- CAS Number:110-83-8
- Formula:C₆H₁₀
- MDL Number:MFCD00001539
- Molecular Weight:82.15 g/mol
- Merck Index:14,2727
- Boiling Pt:83…85 °C
- Melting Pt:–104 °C
- Density:0.811
- Flash Pt:–20 °C (–4 °F)
- Storage Temperature:Ambient
- EINECS:203-807-8
- UN:2256
- ADR:3,II



