Order Entry
Export
ContactUsLinkComponent
Isopropylcyclohexane
Isopropylcyclohexane
Catalog # EHERC14463600
Supplier:  EHRENSTORFER
CAS Number:  
Isopropylcyclohexane
Catalog # EHERC14463600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :126.24 g/mol
Boiling Pt:155 °C (1013 hPa)
Melting Pt:–90 °C
Density:0,8036 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-792-4
UN:3295
ADR:3,I
CAS Number:696-29-7
Formula:C₉H₁₈
MDL Number:MFCD00001480