3-Methyl-1,2-cyclopentanedione ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:765-70-8
:
Formula:C₆H₈O₂
MDL Number:MFCD00001417
:112.13 g/mol
Boiling Pt:210 °C (1013 hPa)
Melting Pt:103…107 °C
Storage Temperature:Ambient
EINECS:212-154-8