3-Methyl-1,2-cyclopentanedione 99%
:765-70-8
:
:C₆H₈O₂
:MFCD00001417
:112.13 g/mol
:210 °C (1013 hPa)
:105…110 °C
:100 °C
:Ambient
:212-154-8
- Appearance:White to yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:104 to 108 °C
- GC:≥98.5 %