2-Methyl-1,3-cyclopentanedione
Catalog # MOLE29962625-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:112.13 g/mol
Boiling Pt:213 °C (1013 hPa)
Density:1.33 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-153-2
CAS Number:765-69-5
Formula:C₆H₈O₂
MDL Number:MFCD00001406