
2-Methyl-1,3-cyclohexanedione 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1193-55-1
:
Formula:C₇H₁₀O₂
MDL Number:MFCD00001587
Boiling Pt:213 °C (1013 hPa)
Melting Pt:>204 °C
Density:1,33 g/cm³ (20 °C)
:126.16 g/mol
Storage Temperature:Ambient
EINECS:212-153-2
:GHS07
Specification Test Results
- Appearance:Off-white to pink-orange to light brown Powder or crystals
- Infrared spectrum:Conforms
- Melting point:≥204 °C
- Water:≤1 % (K.F.)
- Titration with NaOH:≥98.0 %