Order Entry
Export
ContactUsLinkComponent
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Sigma-Aldrich®
Irritant
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Sigma-Aldrich®
Catalog # SIALT5691-500G
Supplier:  Merck
CAS Number:  
Irritant
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Sigma-Aldrich®
Catalog # SIALT5691-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :229.25 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:230-906-3
 :GHS07
Storage Temperature:Ambient
CAS Number:7365-44-8
Formula:C₆H₁₅NO₆S
MDL Number:MFCD00007532